Learn More
Thermo Scientific™ Kinase buffer I (5X)
For cellular and molecular biology applications
Brand:  Thermo Scientific™ J61967.AE
 View more versions of this product
Product Code. 15434449
  • 45.53 € / 100mL
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Formulation:100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4.
100 mL
Liquid
Ambient temperatures
Kinase buffer I (5X)