Learn More
Kinase Buffer R (5X)
Kinase Buffer R (5X)
Brand:  Invitrogen PV5101
Additional Details : Weight : 0.99000kg
Product Code. 10528170
  • 187.00 € / Each
Estimated Shipment: 23-07-2024
to see stock.
Description
Description
Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.
Specifications
Specifications
5X
Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, Lanthascreen™ kinase activity assays, Adapta™ Universal kinase assays, enzyme and
4 mL