missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
418.00 € - 651.00 €
Product Code | Brand | Model | Dimensions (Dia. x H) | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Dimensions (Dia. x H) | Capacity (Metric) | Price | Quantity & Availability | |||
10769754
|
HMC Europe
50008 |
Stackable basket | 390 x 290 mm | 85 L |
418.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10348082
|
HMC Europe
50029 |
With collecting pan | 390 x 290 mm | 85 L |
651.00 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||
Description
For use with the HMC-HG and HMC-HV autoclaves.Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title