Learn More
Kinematica™ H-Foot Stand ST-F 10/600 for Polytron™ PT 3100 D
An H-foot stand made of anodized aluminum.
Brand:  Kinematica™ 11040025
Product Code. 10495342
  • 2125.00 € / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Height adjustable
  • Swiveling PTFE-coated work table with vessel holder
  • 5.5kg in weight
Specifications
Specifications
Homogenizer Stand
Anodized Aluminum
H-foot homogenizer stand
ST-F 10/600