missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, 99%, Thermo Scientific Chemicals

39.70 € - 584.91 €

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.216
MDL Number MFCD00075757
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464724
View Documents
Thermo Scientific Alfa Aesar
A13720.03
1 g
39.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11474724
View Documents
Thermo Scientific Alfa Aesar
A13720.06
5 g
150.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
11484724
View Documents
Thermo Scientific Alfa Aesar
A13720.14
25 g
584.91 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.

Solubility
Soluble in water (<1mg/mL) at 25°C, Acetic acid (49.00-51.00 mg/ml), DMSO (43 mg/ml) at 25°C, ethanol (<1mg/mL) at 25°C, 0.1 N Sodium hydroxide (1% w/v), methanol (slightly), and dilute aqueous hydrochloric acid or sodium hydroxide (freely).

Notes
Protect from heat. Store at -20°C. Store away from oxidizing agents.
Specifications

Chemical Identifiers

525-79-1
215.216
QANMHLXAZMSUEX-UHFFFAOYSA-N
3830
N-(furan-2-ylmethyl)-7H-purin-6-amine
C10H9N5O
MFCD00075757
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
CHEBI:27407
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3

Specifications

525-79-1
99%
MFCD00075757
14,5311
Soluble in water (<1mg/ml) at 25°C,Acetic acid (49.00-51.00mg/ml),DMSO (43mg/ml) at 25°C,ethanol (<1mg/ml) at 25°C,0.1 N Sodium hydroxide (1% w/v),methanol (slightly),and dilute aqueous hydrochloric acid or sodium hydroxide (freely).
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3
215.216
CHEBI:27407
99%
∽270°C (decomposition)
C10H9N5O
21703
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
QANMHLXAZMSUEX-UHFFFAOYSA-N
N-(furan-2-ylmethyl)-7H-purin-6-amine
3830
215.22
Kinetin
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-382-2

missing translation for 'rtecsNumber' : AU6270000

missing translation for 'tsca' : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.