Learn More
Thermo Scientific™ KingFisher™ Apex Dx Magnetic Heads
The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids from human specimens with a small, compact footprint making it ideal for use in diagnostic and development workflows.
Brand:  Thermo Scientific™ B24079940
 View more versions of this product
Product Code. 17846588
  • 7010.00 € / Each
Explore more special offers
Description
Description
KingFisher Apex Dx Magnetic Heads are for use with the KingFisher Apex Dx Purification Instrument. The instrument accommodates two interchangeable magnetic heads (it comes with one included). The interchangeable heads and heating blocks provide flexibility in the extraction of nucleic acids, proteins, and cells with a small, compact footprint, making the instrument ideal for use in diagnostic and development workflows.
With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, the KingFisher Apex Dx Purification Instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nucleic Acid Extraction and Cell Separation Instrument
Apex Dx 24 Combi Head
1 head
Viral Samples
Reusable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number294024010085 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog NumberB24079940
Lot Number294024060112 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog NumberB24079940
Lot Number294024030103 Certificate TypeCertificate of Compliance Date26/01/2025 Catalog NumberB24079940
Lot Number294024030105 Certificate TypeCertificate of Compliance Date26/01/2025 Catalog NumberB24079940
Lot Number294024030102 Certificate TypeCertificate of Compliance Date26/01/2025 Catalog NumberB24079940
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Apex Dx Magnetic Heads > 24 Combi Head
For in vitro diagnostic use.