Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Product Code. 16882406 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
KF Duo Prime, Flex and Presto
KingFisher Apex
KingFisher Duo Prime only
KingFisher Flex and Presto
KingFisher Flex, Duo Prime
Unit Size:
Each
Pack of 100
Pack of 40
Pack of 50
Product Type:
Barcoded Tip Comb
Combi Pack
Elution Strip
Elution Strip Cap
Tip Comb
Tip Comb and Plate
Product Code. 16882406
Brand: Thermo Scientific™ 95040455
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Deep-well Plate
For Use With (Equipment) KingFisher Flex, Duo Prime
No. of Wells 96
Quantity 50/Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.