Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
Brand:  Thermo Scientific™ N12459
 View more versions of this product
Product Code. 30131189
  • 521.00 € / Each
Explore more special offers
Description
Description
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nucleic Acid Extraction and Cell Separation Instrument
50 to 1000 μL
12 Samples
KingFisher Duo Prime Purification System
Fresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma
96 Deep-well Head
Reusable
1 Head