Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
Brand:  Thermo Scientific™ N12460
 View more versions of this product
Product Code. 30132313
  • 536.00 € / Each
Explore more special offers
Description
Description
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nucleic Acid Extraction and Cell Separation Instrument
200 to 5000 μL
6 Samples
KingFisher Duo Prime Purification System
Fresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma
24 Deep-well Head
Reusable
1 Head