missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ KingFisher Duo Prime Accessory Kit B
Click to view available options
:
No. of Reactions: 8 x 6
Description
- Adapted NucleoMag™ procedure for instruments of the KingFisher™ Duo series
- Reliable performance and excellent yields from different sample materials
- Speeds up nucleic acid purification workflow: process up to 12 samples at once
- Set includes Deep-well Plates and Tip Combs for processing
Specifications
Specifications
| Format | Accessory Kit |
| For Use With (Equipment) | Macherey-Nagel™ NucleoMag™ Kits on KingFisher Duo Prime instrument |
| For Use With (Application) | Automated Nucleic Acid Purification |
| No. of Reactions | 8 x 6 |
| Packaging Type | 1 Set |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction