missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex 24 Deep Well heating block

Product Code. 12535977 Shop All Thermo Scientific Products
Click to view available options
Quantity:
Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12535977

Brand: Thermo Scientific™ 24075440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The KingFisher™ Flex 24 Deep-Well Heating Block is compatible with the KingFisher™ Flex 24 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 24 Deep-Well Head.

The KingFisher™ Flex 24 Deep-Well Heating Block is compatible with the KingFisher™ Flex 24 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 24 Deep-Well Head.
TRUSTED_SUSTAINABILITY

Specifications

Format 24 Deep-well Heating Block
Capacity 24 Samples
For Use With (Equipment) KingFisher™ Flex
Product Line KingFisher
Quantity Each
Shipping Condition Room Temperature

For research use only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.