Learn More
Thermo Scientific™ KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge
The KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge is used with the KingFisher™ PlasmidPro Maxi Processor for fully automated isolation of endotoxin-free plasmid DNA in under 75 minutes.
Brand:  Thermo Scientific™ A54072
Product Code. 30131894
  • 495.00 € / Pack of 4
Explore more special offers
Description
Description
The KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge is used with the KingFisher™ PlasmidPro Maxi Processor for fully automated isolation of endotoxin-free plasmid DNA in under 75 minutes. Purify high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.
Features and benefits
  • Convenient one-piece cartridge pre-filled with all necessary reagents
  • Collection tube included for simple removal and storage of plasmid DNA
  • Designed for mess-free clean-up and waste disposal
  • Plasmid DNA contains less than <0.1 EU/μg
Additional product details
The KingFisher™ PlasmidPro Maxi Processor is a cutting-edge solution designed for fully automated maxi-scale plasmid purification.
At the end of the procedure, the system delivers endotoxin-free plasmid DNA. The purified DNA is resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Cartridge
Plasmid purification
Plasmid DNA
<75 min.
Room Temperature
4 KingFisher™ PlasmidPro Maxi Processor Endotoxin-free Cartridges
Store at room temperature (15°C to 25°C).
KingFisher™ PlasmidPro Maxi Processor
4 Preps
150 mL
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato
For Research Use Only. Not for use in diagnostic procedures.