Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
176.00 € - 544.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16694342
|
Thermo Scientific™
A48305 |
KingFisher 96 deep-well plate, v-bottom, polypropylene (for Duo Prime, Flex, Apex and Presto) | N/A | ||||||
16614352
|
Thermo Scientific™
A48438 |
KingFisher Deep Well 96 Tip Comb, 100 PCS | N/A | ||||||
17153249
|
Thermo Scientific™
A48438 |
KingFisher Deep Well 96 Tip Comb, 100 PCS | N/A | ||||||
17102959
|
Thermo Scientific™
267600 |
Tip Comb Presenting Plate for Flex 96 (50 PCS) | N/A | ||||||
Description
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
Optimize performance using Thermo Scientific KingFisher Systems Consumables, designed specifically for use for COVID-19 testing with these instruments. KingFisher instruments utilize disposable plastics made of polypropylene. The plastics, tip combs and microplates, are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads. Specifically tested with the TaqPath COVID-19 Combo Kit and MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.