Learn More
Thermo Scientific™ KingFisher™ Pure DNA Plant Kit
Achieve rapid automated and reproducible purification of high-quality DNA from various fresh or dried plant materials with this unique nucleic acid purification workflow.
Brand: Thermo Scientific™ 98050196
This item is not returnable.
View return policy
Description
Achieve rapid automated and reproducible purification of high-quality DNA from various fresh or dried plant materials using the Thermo Scientific™ KingFisher™ Pure DNA Plant Kit, together with the KingFisher magnetic particle processor. Thermo Scientific KingFisher Kits complete the unique nucleic acid purification workflow providing an optimized high-throughput method for extreme flexibility.
Features:- Produces DNA that is free of inhibitors and ready for use in many downstream applications
- Easy-to-use protocols facilitate safe sample handling as well as minimal user input maximizing walkaway time
- Excellent performance and consistency of results
- Suitable for fresh and dried plant sample materials
- Flexible throughput from one sample to up to 96 samples per run
- Minimized hands on time increases the efficiency
- Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Specifications
Fresh or Frozen Plant Tissues | |
KingFisher Pure DNA Plant Kit; 96 samples | |
96 Samples | |
Up to 50 mg of fresh plant tissue |
PCR, Real-time PCR, Enzymatic Reactions | |
100µL (Duo) or 150µL (Flex) | |
40 min. |
Safety and Handling
Supplemental information
- MIXTURE LIST-Contains : Sodium lauryl sulfate, Guanidine hydrochloride