Learn More
Thermo Scientific™ KingFisher™ Flex 24 Deep Well head and heating block
Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Purification Systems.
Brand:  Thermo Scientific™ 24074441
Product Code. 12589337
  • 4900.00 € / Each
Description
Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Purification Systems, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.
Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96magnet head.
  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20minutes.
  • Expands sample volume by 10-fold with the 24-well format.
  • Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
  • Ready-made protocols for different type of applications are available.
Recommended for:
  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products