Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
Brand:  Thermo Scientific™ 97002080
 View more versions of this product
Product Code. 12673116
  • 310.00 € / Pack of 50
Explore more special offers
Description
Description
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
  • Specifically designed for use with the KingFisher System
  • Ideal for magnetic particle processing because of their low binding affinity for biomolecules
  • The specially-designed, disposable tip combs and microplates are made of polypropylene
  • This enables excellent recovery of magnetic beads
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 μL
KingFisher System
50/Cs.
Magnetic particle processing
KingFisher plate 100 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2410210 Certificate TypeCertificate of Compliance Date28/08/2024 Catalog Number97002080
Lot Number2410010 Certificate TypeCertificate of Compliance Date28/08/2024 Catalog Number97002080
Lot Number2405210 Certificate TypeCertificate of Compliance Date15/08/2024 Catalog Number97002080
Lot Number2324410 Certificate TypeCertificate of Compliance Date13/09/2023 Catalog Number97002080
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Systems Consumables > KingFisher plate 100uL, 50 pcs/box
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.