Learn More
HUBER KISS 208B Heating Bath Circulator
Heating bath circulators with a stainless steel bath allow the temperature control of internal and external thermoregulation tasks. Items can be directly thermoregulated by placing them in a thermostat bath and the control of externally connected applications is also possible.
Brand:  HUBER 2056.0004.98
Product Code. 15692717
  • 2720.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature of various objects can be controlled by placing them directly within bath
  • Fitted with overtemperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids
  • Pressure/suction pump ensures optimal circulation and temperature homogenity in bath
  • Allows temperature control of externally attached applications using pump adapter
  • KISS temperature controller
  • Simple operation with menu navigation
  • Simultaneous display of setpoint, Tmin, Tmax and actual temperature
  • Status displays for pump, cooling and heating
  • USB and RS232 interfaces
  • Autostart function for power failure
  • 3 colour versions available: grey (standard), blue, red
  • Made from stainless steel
  • Suitable for routine tasks in research and industry
Specifications
Specifications
KISS
III/FL (DIN 12876)
25°C to 200°C
2 kW
14 L/min; 0.25 bar
±0.05°C
22 lb.
230 V
50/60 Hz
8.5 L
M16x1 AG Male
KISS 208 B Bath Thermostat with Stainless Steel Bath
Stainless Steel
KISS 208 B
350 mm
10 kg
10 A