missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Copper Catalyst Tablets, Technical, Contains 5g K2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™

Product Code. 10447260
Click to view available options
1000tablets, Plastic powder jar

Kjeldahl Copper Catalyst Tablets, Technical, Contains 5g K2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Blue-White, Quantity: 1000 tablets, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid

TRUSTED_SUSTAINABILITY

Specifications

Color Blue-White
Packaging Plastic powder jar
MDL Number 147336
Quantity 1000 Tablets
Physical Form Solid
Product Identifier
  • Kjeldahl catalyst tablets Copper
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains :
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.