missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Kjeldahl Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
547.00 € - 1090.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10315361
|
Lenz Laborglasintrumente
03052949 |
250 mL |
698.00 €
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
10264621
|
Lenz Laborglasintrumente
03051937 |
100 mL |
570.00 €
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
10420323
|
Lenz Laborglasintrumente
03052964 |
750 mL |
885.00 €
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
10570112
|
Lenz Laborglasintrumente
03051928 |
50 mL |
547.00 €
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
10604392
|
Lenz Laborglasintrumente
03052970 |
1 L |
1090.00 €
Pack of 10 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
Description
- For the determination of nitrogen in organic compounds
- With center neck
- With conical joint and inscription label
- Material: DURAN™
Specifications
Borosilicate Glass (3.3) | |
10/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasintrumente Kjeldahl Flasks