missing translation for 'onlineSavingsMsg'
Learn More
Kjeldahl Reagent 30%, Reagecon™ Product Code.: 11934357
compliance-icons

Kjeldahl Reagent 30%, Reagecon™
BIOPHARMA

Product Code. 11934357 Shop All Reagecon Products
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 11934357

Brand: Reagecon S30WW5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 5 L
Concentration or Composition (by Analyte or Components) 30%

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS30WW524K1 Certificate TypeCertificate of Compliance Date06/01/2025 Catalog NumberS30WW5
Lot NumberS30WW524H1 Certificate TypeCertificate of Compliance Date18/09/2024 Catalog NumberS30WW5
Lot NumberS30WW524A1 Certificate TypeCertificate of Compliance Date04/04/2024 Catalog NumberS30WW5
Lot NumberS30WW523L1 Certificate TypeCertificate of Compliance Date24/01/2024 Catalog NumberS30WW5
Lot NumberS30WW523D1 Certificate TypeCertificate of Compliance Date01/08/2023 Catalog NumberS30WW5
compliance-icons
Product Identifier
  • SODIUM HYDROXIDE 30% w/w
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Sodium Hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kjeldahl Reagent 30%, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.