missing translation for 'onlineSavingsMsg'
Learn More
Kjeldahl Reagent 4%, Reagecon™ Product Code.: 11924357

Kjeldahl Reagent 4%, Reagecon™

Product Code. 11924357 Shop All Reagecon Products
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 11924357

Brand: Reagecon BOA4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kjeldahl Reagent 4%, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.