Learn More
KjTabs VS - Kjeldahl Antifoam Tablets, Velp Scientifica™
KjTabsTM VS Antifoam are perfect to reduce foam formation during the initial phase of the mineralization process.
Brand:  Velp Scientifica A00000283
Product Code. 15581775
  • 269.00 € / Pack of 1000
Explore more special offers
Description
Description
The right KjTabs™ to fit your needs
The KjTabs™ VS Antifoam are specific antifoaming tablets consisting of Potassium sulphate and Silicone.
Antifoaming agents are fundamental to the good outcome of the Kjeldahl analysis since the production of foam tends to reduce the original quantity of the sample during the initial phase of the mineralization process. This can occur in many agricultural samples or fat-rich products.
VS Antifoam used for foam reduction
  • 0,97 g K2SO4 + 0,03g Silicone
  • 1000 pcs
VELP analytical experts KjTabs™ antifoaming tablets application suggestions
The KjTabs™ VS Antifoam are added in combination with KjTabs™ catalylist tablets. The most suitable choice when performing Kjeldahl analysis in a great variety of industries.
Specifications
Specifications
1000 Pc./Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KjTabs VS - Kjeldahl Antifoam Tablets, Velp Scientifica™ > Quantity: 1000 pcs/box