Learn More
KjTabs VST - Kjeldahl Catalyst Tablets, Velp Scientifica™
KjTabsTM VST Kjeldahl Catalyst Tablets are ideal for oily and fatty samples. The perfect solutions to speed up the digestion process.
Brand: Velp Scientifica A00000277
This item is not returnable.
View return policy
Description
The right KjTabs™ to fit your needs
The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses. Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with VELP Scientifica genuine catalyst tablets.The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
The KjTabsTM VST consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Selenium (Se) as metal salt to accelerate the mineralization process.
VST recommended for oily/fatty samples & even faster digestion
3,5g K2SO4 + 3,5mg Se
1000 pcs
VELP analytical experts KjTabs™ catalyst tablets application suggestions
The KjTabs™ VST are the most suitable choice when performing Kjeldahl analysis in several industries such as beverage, agriculture, environmental, chemical & petrochemical:- Protein determination in beer and malt
- Nitrogen determination in soil, plants, forage, straw and fertilizers
- Nitrogen determination in engine oils, crude oil, lubricating oils and fuel oils
Specifications
1000 Pc./Box |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
KjTabs Kjeldahl Catalyst Tablets
File size: 361.9 kB
Product Content Correction
Product Title
KjTabs VST - Kjeldahl Catalyst Tablets, Velp Scientifica™ > Quantity: 1000 pcs/box
Spot an opportunity for improvement?Share a Content Correction