Learn More
KjTabs VTCT - Kjeldahl Catalyst Tablets, Velp Scientifica™
KjTabsTM VTCT Kjeldahl Catalyst Tablets are ideal for tougher samples. The perfect solutions to speed up the digestion process.
Brand:  Velp Scientifica A00000281
Product Code. 15561775
  • 258.00 € / Pack of 1000
Explore more special offers
Description
Description
The right KjTabs™ to fit your needs
The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses. Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with VELP Scientifica genuine catalyst tablets.
The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
The KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
VTCT for tougher samples & even faster digestion (a scaled down version of VCT)
3,5g K2SO4 + 0,105g CuSO4 x 5 H2O + 0,105g TiO2
1000 pcs
VELP analytical experts KjTabs™ catalyst tablets application suggestions
The KjTabs™ VTCT are the most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
Specifications
1000 Pc./Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KjTabs VTCT - Kjeldahl Catalyst Tablets, Velp Scientifica™ > Quantity: 1000 pcs/box