missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenex™ Extra Gel Hand Sanitiser

Fortified with emollients and kills up to 99.99% of a wide range of microorganisms. With a dye and fragrance free formulation, this alcohol gel is dermatologically tested. The Kleenex™ Extra Gel Hand Sanitiser refills are designed for quick and easy installation with a simple push and click.

152.00 € - 156.00 €

Specifications

Color Clear
Packaging Cassette
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15688740
View Documents
Kimberly-Clark™
6383
4 x 1.2 L
152.00 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
16634472
View Documents
Kimberly-Clark™
6382
6 x 1 L
156.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Designed to deliver an unrivalled experience whilst supporting high hand hygiene standards in the workplace.
  • This alcohol hand sanitiser kills up to 99.99% of a wide range of microorganisms.
  • The hand gel is suitable for use in hygiene-critical areas such as healthcare and food processing environments and meets the requirements of the Biocidal Products Regulation (BPR, Regulation (EU) 528/2012).
  • This Kleenex™ hand sanitiser conforms to EN12791, EN13624, EN13727, EN14476, EN14348, EN1500.
  • The dye and fragrance free formulation of this gel hand sanitiser is dermatologically tested and is fortified with emollients to support frequent use.
  • The hand sanitiser refills are compatible with a range of Aquarius™ hand cleanser dispensers (product codes 6948, 6955, 7124 and 7173).
  • The hand sanitiser gel cassettes are designed for quick and easy installation with a simple push and click.
  • When the alcohol gel cassette is empty, it can be compressed to conserve space and is fully recyclable once the pump is removed.
Specifications

Specifications

Clear
Cassette
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • KLEENEX Gel Hand Sanitizer

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : tetradecanol, ethanol, glycerol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.