missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kleenex™ Ultra™ Paper Towels
Dry your hands hygienically and effectively, and help reduce costs and waste with these paper towels. Kleenex™ Ultra™ Paper Towels are ideal for food contact areas and office and hotel washrooms.
Brand: Kimberly-Clark™ 6778
66.50 EUR valid until 2024-12-31
Use promo code "24047" to get your promotional price.
This item is not returnable.
View return policy
Description
- Interfolded sheets
- Highly absorbent Airflex™ fabric
Specifications
Hand Towel | |
31.5 cm | |
21.5 cm | |
White | |
Food contact areas and office and hotel washrooms that have a high-quality image, providing a luxury feel, drying your hands quickly and comfortably with fewer towels, as they’re large, strong and highly absorbent without breaking up when wet | |
2-ply |
21.81 in. | |
13.1 in. | |
Paper | |
31.5 x 21.5 cm | |
15 Packs x 124 Sheets (1860 Sheets) |