missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ A20+ Breathable Particle Protection Coveralls

Product Code. 13404566
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to protect against particles, fibers, and light sprays. Breathable particle protection coveralls are manufactured from lightweight SMS fabric to aid the body in maintaining thermal comfort.

  • Ergonomically designed body, hood and waist for comfort
  • Thumb loops to help prevent sleeves from riding up
  • Triple-stitched internal seams to help resist general wear and tear and fabric stain
  • Silicone-free and anti-static coated for when working in critical areas like painting or sensitive surfaces
  • Outer abrasion-resistant layer for durability, especially when crawling or rubbing against objects like walls, etc.

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Stitched
Sterility Sterile
Resistance Tears
Back Style Stretchable
Certifications/Compliance (Europe) EN 1073-2:2002, EN1149-1, CE mark, Type 5, Type 6
Latex Free Yes
For Use With (Application) General Maintenance, Contract Cleaning, Construction Utility
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'