Learn More
Ecolab™ Klercide™ 70/30 Denatured Ethanol Blended With WFI
70% denatured ethanol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
Brand:  Ecolab™ 3078340
Product Code. 15396068
  • 913.00 € / Pack of 4
Explore more special offers
Description
Description
  • The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
Specifications
Specifications
4 Pk.
5 L