missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab Klercide 70/30 IPA Blended with Deionized Water
Klercide 70/30 IPA blended with deionized water is sterile and 0.2 micron filtered.
718.00 € - 1500.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17825497
|
Ecolab
6600967 |
12 x 500 mL |
718.00 €
6 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17835497
|
Ecolab
6600956 |
4 x 5 L |
1500.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17805497
|
Ecolab
6600946 |
6 x 1 L |
928.00 €
6 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- BPR approved
- Economical, high grade disinfection with easy application
- 0.2 micron filtered, filled and triple bagged in a Grade C (ISO Class 7) cleanroom
- Irradiated at no less than 25 kGy to ensure a sterile alcohol
- Supplied in a trigger spray incorporating the Sterishield Delivery System which ensures in-use sterility for up to 3 months
- The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use
- Certificates of Analysis are available for every batch
- Full supporting technical documentation is available
Specifications
Disinfectant | |
Liquid | |
Neutral | |
Sterile |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 70/30 IPA
Signal Word
- Danger
Hazard Category
- Flammable and non-flammable aerosols category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H222-Extremely flammable aerosol.
- H229-Pressurised container: May burst if heated.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P211-Do not spray on an open flame or other ignition source.
- P251-Do not pierce or burn, even after use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P410-Protect from sunlight.
- P412-Do not expose to temperatures exceeding 50°C/122°F.
Supplemental information
- MIXTURE LIST-Contains : Propan-2-ol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title