missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klercide 70/30 IPA Blended with Water for Injection

Klercide 70/30 IPA blended with the highest grade (EP) Water for Injection providing a versatile, high quality product for use in Grade A and B cleanrooms.

1690.00 €

Specifications

Product Type Disinfectant
Form Liquid
pH Neutral
Sterility Sterile
View More Specs

Products 1
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17855497
View Documents
Ecolab
6600944
6 x 1 L
1690.00 €
6 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • BPR approved
  • Versatile, high grade disinfection with easy application
  • Triple bagged for easy transfer into Grade A and B cleanrooms
  • 0.2 micron filtered, filled and triple bagged in a Grade C (ISO Class 7) cleanroom
  • Irradiated at no less than 25 kGy to ensure a sterile alcohol
  • Endotoxin tested to an approved pharmacopoeial method
  • Supplied in a trigger spray incorporating the Sterishield Delivery System which ensures in-use sterility for up to 3 months
  • The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available
Specifications

Specifications

Disinfectant
Liquid
Neutral
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.