missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide Sporicidal Enhanced Peroxide Blended with DI Capped
Klercide Sporicidal Enhanced Peroxide is an enhanced blend of 1.5% hydrogen peroxide with deionized water that is sterile and ready-to-use. This biocide blend is 0.2 μm filtered then sterile filled and double bagged under Grade A laminar flow.
Brand: Ecolab™ 6600525
This item is not returnable.
View return policy
Description
- BPR approved
- Ready-to-use disinfectant for proactive control of spores, particularly on larger surfaces where safe handling is especially important
- Short contact times against bacteria and fungi even in the presence of background soiling
- Low odor: Safe for operators and cleanroom surfaces
- Effective broad spectrum disinfectant efficacy even when the level of bioburden may be variable, such as during transfer disinfection
- No hazard classification: Free of aldehyde, chorine and peracetic acid
- Broad virucidal efficacy
- 5 L canister for large surface mopping
- Double bagged
- Low surface tension properties which ensure an optimum spread of the disinfectant and ideal surface wetting for effective results
- Certificates of Analysis and Sterility are available for every batch
- Full supporting technical documentation is available
Specifications
Cleanroom Biocide | |
2.01 to 2.30 | |
Sterile | |
1.5% Hydrogen Peroxide with Deionized Water | |
Single-use | |
5 L |
Liquid | |
Unscented | |
Clear to Light Yellow | |
Capped Bottle | |
No | |
5°C to 25°C |