missing translation for 'onlineSavingsMsg'
Learn More

IKA™ KMO 3 Basic Magnetic Stirrer
GREENER_CHOICE

A small but powerful magnetic stirrer without heating function.

Brand:  IKA™ 0020004631

738.15 EUR valid until 2025-12-31
Use promo code "24299" to get your promotional price.


Product Code. 15719394

  • 777.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unique design with glass surface
  • Brilliant, clear white display
  • Illuminated, flat stirring plate
  • Key-lock button
  • Reverse rotation
  • Integrated USB interface (control via labworldsoft and use of Firmware Update Tool)
  • Recessed thread for support rod
  • Protection class IP42
Specifications

Specifications

10 to 1500 rpm
Glass
LED
0.8 W
140 mm
50/60 Hz
Touch Key
1
5 L
100/240 V
227 x 153 x 62.5 mm
120 mm
UK-Style
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.