missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KN-93, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Potent, cell permeable inhibitor of CaM kinase II (IC50 = 0.37 μM). Also a direct extracellular open channel blocker of voltage-gated potassium channels (IC50 = 307 nM for KV1.5) and abolishes IKr in ventricular myocytes (IC50 = 102.6 nM) independently of CaM kinase II inhibition.
Water soluble form and Negative Control also available.
Specifications
Specifications
CAS | 139298-40-1 |
Molecular Formula | C26H29ClN2O4S |
Quantity | 1 mg |
Synonym | methoxybenzene-sulfonamide, insolution kn-93, e-n-2-3-4-chlorophenyl allyl methyl amino methyl phenyl-n-2-hydroxyethyl-4-methoxybenzenesulfonamide, 2-n-2-hydroxyethyl-n-4-methoxybenzenesulfonyl amino-n-4-chlorocinnamyl-n-methylbenzylamine, benzenesulfonamide, n-2-3-4-chlorophenyl-2-propenyl methylamino methyl phenyl-n-2-hydroxyethyl-4-methoxy, n-2-3-4-chlorophenyl-2-propenyl methylamino methyl phenyl-n-2-hydroxyethyl-4-methoxybenzenesulfonamide, n-2-2e-3-4-chlorophenyl prop-2-en-1-yl methyl amino methyl phenyl-n-2-hydroxyethyl-4-methoxybenzenesulfonamide, kn-93 free base |
InChI Key | LLLQTDSSHZREGW-AATRIKPKSA-N |
SMILES | CN(CC=CC1=CC=C(C=C1)Cl)CC2=CC=CC=C2N(CCO)S(=O)(=O)C3=CC=C(C=C3)OC |
IUPAC Name | N-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-N-(2-hydroxyethyl)-4-methoxybenzenesulfonamide |
Molecular Weight (g/mol) | 501.038 |
PubChem CID | 5312122 |
Formula Weight | 501.04 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction