missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ KnockOut™ Serum Replacement

KnockOut™ Serum Replacement
106.00 € - 473.00 €
Specifications
Classification | Serum-Free |
---|---|
Form | Frozen |
Serum Level | Serum-Free |
Product Type | Serum Replacement |
Product Line | KnockOut™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10829220
![]() |
Gibco™
10828010 |
100 mL |
106.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11520366
![]() |
Gibco™
10828028 |
500 mL |
473.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ KnockOut™ Serum Replacement (KnockOut™ SR) is a more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells. Designed to replace fetal bovine serum (FBS) in existing protocols, this serum replacement has been trusted by many for almost 20 years. Providing the documents required for regulatory review, KnockOut SR reduces your burden in qualifying reagents when transitioning from research applications to clinical applications. If you are interested in only basic research applications, see the Research Use Only version of this product to ensure that you receive the benefits and pricing that fit your needs.With KnockOut Serum Replacement, you can:
Culture PSCs with confidence
KnockOut SR has been cited in more than 2000 publications. It has been shown to support feeder-dependent culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs). It has also been shown to support reprogramming, embryoid body (EB) formation, in vitro differentiation, cryopreservation, ESC derivation, and many other applications.
Rely on improved performance
KnockOut SR has more a defined formulation compared to FBS, which contains many undefined and variable components. Unlike most FBS products, each lot is tested for its ability to support PSCs. Because of this, culturing PSCs with KnockOut SR leads to less differentiation and more compact colony morphology compared to FBS.
Enjoy the peace of mind of having stable price and supply
KnockOut SR is not affected by the limited supply and fluctuating prices often seen with FBS. This makes it easier to plan your budget and buy what you need, especially since it is more affordable than most FBS products.
This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in a specific region, it should be used for laboratory use only.
Specifications
Serum-Free | |
Serum-Free | |
KnockOut™ |
Frozen | |
Serum Replacement | |
Dry Ice |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ KnockOut™ Serum Replacement