missing translation for 'onlineSavingsMsg'
Learn More

Koflo 316 Stainless Steel In-Line Pipe Mixer

Mix fluids as you pump them through a pipe line with this 316 Stainless Steel In-Line Pipe Mixer.

Brand:  Koflo 04669-16

 View more versions of this product

Product Code. 11724119

  • 365.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Offers 100% mixed output.
  • Industrial grade materials ensure long service life.
  • Durability under demanding conditions.
  • Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe.
  • All mixers have threaded ends.
  • Choose PVC, 316 stainless steel, PVDF, or PFA.
  • The clear PVC mixers are ideal for a visual check of your mixing processes.
  • Models with removable elements are available on request.
Specifications

Specifications

1.1 in.
9 in.
Stainless Steel (316)
Mixing
26.7 mm
229 mm
Pipe Mixer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers