missing translation for 'onlineSavingsMsg'
Learn More

Kojic acid, 99%, Thermo Scientific Chemicals

44.22 € - 1383.89 €

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00006580
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
Synonym kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
PubChem CID 3840
ChEBI CHEBI:43572
IUPAC Name 5-hydroxy-2-(hydroxymethyl)pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10105030
View Documents
Thermo Scientific Acros
205220050
5 g Glass bottle
44.22 €
5g
Please to purchase this item. Need a web account? Register with us today!
10656432
View Documents
Thermo Scientific Acros
205220250
25 g Glass bottle
135.68 €
25g
Please to purchase this item. Need a web account? Register with us today!
10666432
View Documents
Thermo Scientific Acros
205221000
100 g Glass bottle
480.39 €
100g
Please to purchase this item. Need a web account? Register with us today!
11936911
View Documents
Thermo Scientific Acros
205225000
500 g Glass bottle
1383.89 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

501-30-4
142.11
BEJNERDRQOWKJM-UHFFFAOYSA-N
3840
5-hydroxy-2-(hydroxymethyl)pyran-4-one
C6H6O4
MFCD00006580
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
CHEBI:43572
OCC1=CC(=O)C(O)=CO1

Specifications

501-30-4
Authentic
C6H6O4
18,II,57
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
BEJNERDRQOWKJM-UHFFFAOYSA-N
5-hydroxy-2-(hydroxymethyl)pyran-4-one
3840
142.11
Crystalline Powder
151.0°C to 156.0°C
99%
MFCD00006580
15,5365
Solubility in water: soluble. Other solubilities: freely soluble in acetone and ethanol;,sparingly soluble in diethyl ether,ethyl acetate,,chloroform and pyridine;,insoluble in benezne
OCC1=CC(=O)C(O)=CO1
142.11
CHEBI:43572
99%
Kojic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 207-922-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.