Learn More
DWK Life Sciences Kimble™ Kontes™ Airless-Ware Glass Plug Gas Inlet Adapter
Inlet Adapter for Vacuum and AIRLESS-WARE™, with a choice of 14/20 or 24/40 standard taper joints.
Brand:  DWK Life Sciences 211300-1420
 View more versions of this product
Product Code. 15253886
Description
Description
  • Inner Standard Taper joints
  • 211300 series has a glass plug
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
98mm
Borosilicate Glass
14/20 Inner
60mm
3.85 in.
Patented 809000 pressure/vacuum retainer
Inlet Adapter
2.36 in.