Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column PEEK Unions
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
Brand: DWK Life Sciences 420818-3305
This item is not returnable.
View return policy
Description
- EFTE Flangeless Ferrule for 1.8mm OD Tubing
- for Use with 420821 Series Male Nuts
- CHROMAFLEX™ FITTINGS
- Union connects tubing and end fittings F1/4-28 to F1/4-28
- PEEK Internally Threaded Union for use with flangeless fittings
- Case of 1
- Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Flangeless fitting | |
Chromatography Union | |
PEEK | |
F1/4-28 to F5/16-24 |