missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers

Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.

58.29 € - 212.00 €

Specifications

Color White
Material Rubber
Product Type Plug-type Sleeve Stopper
For Use With (Equipment) NMR tubes, pressure venting or adding reactants via syringe
View More Specs

Products 3
Product Code Brand Diameter (Metric) Standard Taper Joint Price Quantity & Availability  
Product Code Brand Diameter (Metric) Standard Taper Joint Price Quantity & Availability  
12859518
View Documents
DWK Life Sciences
774261-0014
1.4 cm 14/20 N/A N/A N/A
15857691
View Documents
DWK Life Sciences
774261-0014
1.4 cm 14/20
58.29 €
Pack of 50
Estimated Shipment: 10-11-2025
to see stock.
Add to basket
 
12869518
View Documents
DWK Life Sciences
774261-0024
2.4 cm 24/40; 24/20
212.00 €
Pack of 50
Estimated Shipment: 10-11-2025
to see stock.
Add to basket
 
Description

Description

Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.

  • Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
  • Diaphragm can be punctured with a syringe needle
  • Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
Specifications

Specifications

White
Rubber
Plug-type Sleeve Stopper
NMR tubes, pressure venting or adding reactants via syringe
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.