Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE Tubing
Chemically inert; suitable for low pressure applications
Brand:  DWK Life Sciences 420823-0116
Product Code. 15370158
  • 69.60 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Chemically inert; suitable for low pressure applications
  • 1/16 OD x 1/32 in. ID x 10 ft. L
  • Outer diameter tolerances of the 1/16 in. O.D. tubing is held at ±0.001 in., assuring a good connection with flangeless nuts and ferrules
  • Tubing designed for easy connection to Flex-Columns™
  •  
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/32 in.
1/16 in.
Suitable for low pressure applications
PTFE
10 ft.