Learn More
VELP Scientifica™ KS 1000 Scrubber
BIOPHARMA
Designed to neutralize toxic fumes produced during mineralization and safeguard laboratory operators without requiring any connection to tap water.
Brand:  VELP Scientifica™ F307A0660
Product Code. 17804687
  • 2925.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Innovative and compact device that is designed to neutralize toxic fumes produced during mineralization and safeguard lab operators, without requiring any connection to tap water.
  • Perfectly complements VELP's compact semi-automatic and automatic digesters, fitting seamlessly into the fume hood for an optimized working environment.
  • An intuitive user interface, conveniently placed on top of the instrument, simplifies operations.
  • With 4 adjustable suction power levels, laboratory operators have control over their specific requirements.
  • Carefully selected materials ensure durability even in the most demanding conditions with reduced maintenance costs.
  • Tanks made of glass and connection tubes constructed with Viton offer superior performance and increased resistance.
  • The eight punctured discs contribute significantly to achieving maximum neutralization efficiency.
  • The innovative design allows for easy access to all components, facilitating quick and effortless cleaning and maintenance.
  • In specific applications, an optional activated carbon filter can be used to absorb odours produced during oxidative mineralization.
Specifications
Specifications
Fume Removal System
3.5 L
KS 1000 Scrubber
No Display
Up to 25 L/min.
Metal structure and PVC components