Learn More
Heidolph™ Kugelmeiers 3D Cell Culture Plate SP5D (Sphericalplate 5D)
The spherical plate 5D (SP5D) is a 3D cell culture plate for the formation of uniform, standardized spheroids with high quality and yield. It allows the effortless scaling and translation into the diagnostic or clinical application.
Brand:  Heidolph™ 549-16000-00
 View more versions of this product
Product Code. 17808808
  • 143.00 € / Each
Explore more special offers
Description
Description
The patented geometry with special coating allows the integration of every single cell into a 3D-agglomerate in a controlled way. This is how uniformly sized spheroids grow without the formation of necrotic cores. The reproducibility of follow-up tests increases, since the experiments always start under identical initial conditions due to the low size variance of the cultivated spheroids.
  • According to the size and scale of the experiments, you can choose between our 24-well plate or our 6-well plate
  • For small batches, Heidolph™ suggests using the 24-well plate containing 12 wells with 750 microwells each so you will be able to yield up to 9000 uniform spheroids
  • You can comfortably carry out monolayer comparative studies in the remaining 12 wells of the same plate
  • 6-well plate is suitable for scale-up trials and bigger batches; you can expect a yield of 3364 spheroids per well, thus a total yield of 20,184 spheroids/plate
  • The ready to use SP5D is particularly user-friendly: The cultivation does not require any pretreatment or centrifugation steps after seeding your cells
  • The media change is also particularly convenient due to simple pipetting, since the height of the microwells was chosen so that the cell clusters are retained
  • Platform is compatible with standard automation units
Specifications
Specifications
No Barcode
With Lid
Not Autoclavable
Single-use
Cycloolefin Copolymer
Sterile
Round, Conical Bottom (V-well)
Plate
Standard
4 x 6
Transparent
Standard Automation Unit
24
Non-binding
Spheroid Cultivation
Sterile & PCR Ready