missing translation for 'onlineSavingsMsg'
Learn More

KV1.3 (KCNA3) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA533863

Product Code. 15322781

  • 470.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Dog, Rabbit, Pig (100%); Bovine, Opossum (94%); Lizard (88%); Turkey, Chicken (81%). Antigen retrieval requires Proteinase K treatment.

In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KV1.3 (KCNA3)
Polyclonal
Unconjugated
KCNA3
HGK5; HLK3; HPCN3; HUK3; HUKIII; Kca1-3; Kcna3; Kv1.3; KV3; MK3; Mk-3; PCN3; potassium channel 3; potassium channel, voltage gated shaker related subfamily A, member 3; potassium voltage gated channel, shaker related subfamily, member 3; potassium voltage-gated channel subfamily A member 3; potassium voltage-gated channel, shaker-related subfamily, member 3; RCK3; RGK5; type n potassium channel; voltage-gated K(+) channel HuKIII; voltage-gated potassium channel protein Kv1.3; Voltage-gated potassium channel protein Kv1.3 (RGK5) (RCK3) (KV3); voltage-gated potassium channel subunit Kv1.3; Voltage-gated potassium channel subunit Kv3
Rabbit
Antigen affinity chromatography
RUO
100156614, 100328945, 100760038, 16491, 29731, 3738
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P15384, P16390, P22001
KCNA3
Synthetic 16 amino acid peptide from C-terminus of human KCNA3 / Kv1.3.
50 μg
Primary
Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.