missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Product Code. 11403737
Click to view available options
Quantity:
1 g
5 g
25 g

L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

Solubility
Solubility in water gives very faint turbidity. 

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20312-36-1
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00004244
InChI Key VOXXWSYKYCBWHO-QMMMGPOBSA-M
Synonym l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
PubChem CID 444718
ChEBI CHEBI:43065
IUPAC Name (2S)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@@H](CC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 122°C to 126°C
Quantity 1 g
Beilstein 2209792
Optical Rotation −24° (c=1 in Water)
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material L-(-)-3-Phenyllactic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'