missing translation for 'onlineSavingsMsg'
Learn More

L-838,417, Tocris Bioscience™

Product Code. 18086012
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GABAA partial agonist; displays subtype selectivity

Subtype-selective GABAA receptor partial agonist. Selectively binds to α1, α2, α3 and α5 subunits (Ki values are 0.79, 0.67, 0.67 and 2.25 nM respectively) but displays no efficacy at α1 (α1-sparing). Exhibits non-sedative anxiolytic, antinociceptive and anti-inflammatory activity in vivo.

Chemical Identifiers

CAS 286456-42-6
Molecular Formula C19H19F2N7O
Molecular Weight (g/mol) 399.41
MDL Number MFCD08690609
InChI Key BQDUNOMMYOKHEP-UHFFFAOYSA-N
PubChem CID 9908880
IUPAC Name 5-({[7-tert-butyl-3-(2,5-difluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazin-6-yl]oxy}methyl)-1-methyl-1H-1,2,4-triazole
SMILES CN1N=CN=C1COC1=NN2C(C=C1C(C)(C)C)=NN=C2C1=C(F)C=CC(F)=C1

Specifications

Quantity 10 mg
Synonym unii-8czo0970g3, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine, 7-tert-butyl-3-2,5-difluorophenyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4 triazolo 4,3-b pyridazine, 5-7-tert-butyl-3-2,5-difluorophenyl-1,2,4 triazolo 4,3-b pyridazin-6-yl oxy methyl-1-methyl-1,2,4-triazole, 1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, l hplc, 1,2,4-triazolo 4,3-b pyridazine,3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-2-methyl-2h-1,2,4-triazol-3-ylmethoxy-1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4,-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine
Formula Weight 399.4
Percent Purity >98%
Chemical Name or Material L-838, 417
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'