missing translation for 'onlineSavingsMsg'
Learn More
L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™ Product Code.: 15459008

L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™

Product Code. 15459008
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15459008

Brand: Thermo Scientific Alfa Aesar H62490.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39825-33-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD08059709,MFCD08059709,MFCD12796134
InChI Key QDQVXVRZVCTVHE-UHFFFAOYNA-N
Synonym s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
PubChem CID 13089460
IUPAC Name propan-2-yl 2-aminopropanoate
SMILES CC(C)OC(=O)C(C)N

Specifications

Quantity 250 mg
Formula Weight 167.63
Percent Purity 95%
Chemical Name or Material L-Alanine isopropyl ester hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.