missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-allo-Threonine, 99%
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass Bottle |
Linear Formula | CH3CH(OH)CH(NH2)CO2H |
Beilstein | 04, III, 1629 |
Specific Rotation | +8° to +10° (20°C, 589nm) (c=2, water) |
Specific Rotation Condition | + 9.00 (20.00°C c=2,H2O) |
Quantity | 250 mg |
Formula Weight | 119.12 |
Show More |
Safety and Handling
Product Identifier
- L(+)-allo-Threonine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals L(+)-allo-Threonine, 99% >
Spot an opportunity for improvement?Share a Content Correction