missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals

Product Code. 10512174
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-89-8
Molecular Formula C40H80NO8P
Molecular Weight (g/mol) 734.053
MDL Number MFCD00036903
InChI Key KILNVBDSWZSGLL-KXQOOQHDSA-N
Synonym colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a
PubChem CID 452110
ChEBI CHEBI:72999
IUPAC Name [(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 2.5 g
Specific Rotation Condition + 7.00 (20.00°C c=2,CHCl3)
Specific Rotation +6.0° to +8.0° (20°C, 589nm) (c=2, CHCl3)
Formula Weight 734.08
Percent Purity ≥97.5% (Fatty acid purity)
Physical Form Powder
Chemical Name or Material L-α-Dipalmitoyl phosphatidylcholine, 98%
Product Identifier
  • L-a-Dipalmitoyl phosphatidylcholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.