missing translation for 'onlineSavingsMsg'
Learn More

L-AP4, Tocris Bioscience™

Product Code. 18005232
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective group III mGlu agonist

Selective group III metabotropic glutamate receptor agonist. Synaptic depressant. Agonist at the quisqualate-sensitized AP6 site in hippocampus.

Available as part of the Group III mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . DL Mixture and D-isomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23052-81-5
Molecular Formula C4H10NO5P
Molecular Weight (g/mol) 183.1
InChI Key DDOQBQRIEWHWBT-VKHMYHEASA-N
Synonym l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid
PubChem CID 179394
IUPAC Name (2S)-2-amino-4-phosphonobutanoic acid
SMILES C(CP(=O)(O)O)C(C(=O)O)N

Specifications

Quantity 1 mg
Formula Weight 183.1
Percent Purity >99%
Chemical Name or Material L-AP4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.