missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Arabinose, 99%, for biochemistry

Product Code. 15289285 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15289285

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O

Specifications

Melting Point 155.0°C to 160.0°C
Ignition Residue 0.3% max
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Quantity 25 g
Beilstein 31,32
Merck Index 10,783
Specific Rotation + 103.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material L(+)-Arabinose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.